Would you like to log in again?
Accucold's General Purpose Fridge-Freezers
Supplier: FELIX STORCH, INC./SUMMIT
Accucold's
AGP34RFCALADA AGP34RFLCAL
470351-066EA 1799 USD InStock
470351-066 470351-064
Accucold's General Purpose Fridge-Freezers
Refrigerators
Digital controls for the refrigerator section with a 2 to 8 °C temperature range and front combination lock to secure both compartments.
  • Interior light -automatically illuminates when the door is opened
  • Thin-line design: slim width and shallow depth mean an easy fit in areas with space constraints
  • Easy operation with a factory-installed digital thermostat (refrigerator) that allows you to manage the refrigerator temperature between 2 and 8 ºF ( freezer temperature will self-adjust based on the refrigerator setting)
  • Alarm/thermometer included- Temperature monitoring device (TMD) displays the current and min/max temperature of the refrigerator interior, an audible alarm alerts if the unit goes out of range
  • Buffered temperature probe - probe is encased in a glycol-filled bottle to better simulate the temperature of stored contents instead of refrigerator air temperature
  • Thermometer is NIST calibrated in ISO/IEC 17025:2017 accredited laboratory and is supplied with a 2-year certificate of calibration for your records
Flexible design to make the most of the interior space, cabinet and doors have a clean white finish. User-reversible swing door for added flexibility. Magnetic door gaskets ensure a tight seal for energy savings and improved performance.
The refrigerator section includes a factory-installed fan to help ensure a more evenly cooled interior. The lower refrigerated section does not require manual defrosting. There is a static cooling system in the upper freezer compartment.
Environmentally friendly design without ozone-damaging chemicals. Environmentally friendly hydrocarbon-based refrigerant for a greener footprint that meets current CARB regulations.
Certifications: ETL.
Delivery information: Supplied with 2 keys to override the combination as needed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty