Would you like to log in again?
Benchtop Centrifuge
Supplier: LW Scientific
M5C-24AU-15T1
470351-006EA 2749 USD InStock
470351-006
Benchtop Centrifuge
Centrifuges Benchtop Centrifuges
A high-performance, versatile large-volume centrifuge.
  • Digital menu is simple-to-use
  • Digital interface ensures accurate processing
  • It spins with microprocessor speed control
  • The wide-mouth metal bowl allows for easy cleanup
The zero-rpm interlocking lid will stay locked until the rotor has come to a stop, to ensure safety. The all-metal rotors are built for strength and durability.
The whisper-quiet heavy-duty brushless DC motor can handle high volume use and high-speed full loads for years to come.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty