Would you like to log in again?
Water Bath
Supplier: LW Scientific
WBL-10LC-SSD1
470351-014EA 1319 USD InStock
470351-014
Water Bath
Baths Water Baths, Stirred
The LW Water Bath creates a constant temperature with an open reservoir and immersion circulator, ideal for heating temperature-sensitive samples. The stainless steel reservoir is seamless, eliminating the possibility of rust and leaks, and the rounded corners are easy to clean.
  • Excellent temperature control
  • Convenient large bath opening
  • Stainless steel reservoir
The control unit achieves the desired temperature quickly and maintains a constant temperature to prevent sample overheating. Select desired setting with the touch-pad digital controller, and view the actual temperature on the red LED display. The compact design and stainless steel reservoir make this product a must for every lab.
Temperature setting: Digital
Temperature readout: LED
Temperature sensor: 100 ohm Platinum RTD
Temperature control: PID
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty