Would you like to log in again?
BD MAX™ PCR Cartridge
Supplier: BD Diagnostics
437519
76808-682CS 421.18 USD InStock
76808-682
BD MAX™ PCR Cartridge
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The BD MAX™ System offers an efficient path to improved clinical outcomes by combining and automating extraction and thermocycling into a single platform capable of running both FDA-cleared and open system assays. Flexibility and standardization allow users to address the breadth of testing needs.
  • BD MAX™ Molecular Diagnostic Testing
  • Use with the BD MAX™ System
  • 24 lane cartridges
BD MAX™ consumables: microfluidic cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty