Would you like to log in again?
Syringe Tips, Corning® Step-R™
Supplier: Corning
Corning® Step-R™
6620 6631 6629 6635 6634 6630 6624 6621 6628 6627 6623 6625 6632 6622 6626 6633
76181-508CS 76.69 USD InStock
76181-508 76181-506 76181-478 76181-488 76181-510 76181-476 76181-486 76181-468 76181-512 76181-492 76181-470 76181-480 76181-490 76181-474 76181-484 76181-482
Syringe Tips, Corning® Step-R™
Pipette Tips
Syringe tips for use with the Step-R™ repeating pipettor. Ideal for viscous, dense and volatile liquides.
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Available sterile or non sterile.
Ordering information: Tip adapters are required for using the 25 mL and 50 mL tips.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty