Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A101478-5G A101478-100G A101478-25G A101478-500G A101478-10G
76809-250EA 8.24 USD InStock
76809-250 76809-252 76809-254 76809-256 76809-248
2,4-Dimethoxybenzaldehyde 98%
2,4-Dimethoxybenzaldehyde
Caution: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00003311
CAS Number: 613-45-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty