Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethyl)aniline 95%

Supplier: Ambeed
Danger

A191488-1G A191488-250MG A191488-100MG
76813-910EA 39.14 USD InStock
76813-910 76813-912
3-(Difluoromethyl)aniline 95%
3-(Difluoromethyl)aniline
Caution: for Research Use Only


Formula: C₇H₇F₂N
MW: 143.137 g/mol
MDL Number: MFCD18384830
CAS Number: 368-99-0
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty