Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-thiourea 97%

Supplier: Ambeed
Danger
Synonyms: tert-Butyl carbamothioylcarbamate

A549497-250MG A549497-100MG A549497-1G A549497-5G
76819-044EA 159.65 USD InStock
76819-044 76819-042 76819-040 76819-038
N-Boc-thiourea 97%
N-Boc-thiourea
Caution: for Research Use Only


Formula: C₆H₁₂N₂O₂S
MW: 176.234 g/mol
MDL Number: MFCD04039987
CAS Number: 268551-65-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty