Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nɑ-Carbobenzoxy-D-arginine 98%

Supplier: Ambeed
Warning
Synonyms: Nɑ-Cbz-D-arginineZ-D-Arg-OH

A694295-5G A694295-10G
76820-006EA 43.26 USD InStock
76820-006 76820-008
Nɑ-Carbobenzoxy-D-arginine 98%
Nalpha-Carbobenzoxy-D-arginine
Caution: for Research Use Only


Formula: C₁₄H₂₀N₄O₄
MW: 308.34 g/mol
MDL Number: MFCD00063009
CAS Number: 6382-93-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty