Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dichloro-6-(trifluoromethyl)benzoic acid 97+%

Supplier: Ambeed
Warning

A726714-1G A726714-250MG A726714-5G
76820-178EA 22.66 USD InStock
76820-178 76820-176 76820-180
2,3-Dichloro-6-(trifluoromethyl)benzoic acid 97+%
2,3-Dichloro-6-(trifluoromethyl)benzoic acid
Caution: for Research Use Only


Formula: C₈H₃Cl₂F₃O₂
MW: 259.01 g/mol
MDL Number: MFCD06660270
CAS Number: 25922-43-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty