Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-2-(5-indolyl)acetic acid 95%

Supplier: Ambeed
Warning
Synonyms: 2-Amino-2-(1H-indol-5-yl)acetic acid

A149293-250MG
76823-780EA 17.51 USD InStock
76823-780
2-Amino-2-(5-indolyl)acetic acid 95%
2-Amino-2-(5-indolyl)acetic acid
Caution: for Research Use Only


Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
MDL Number: MFCD06656882
CAS Number: 108763-43-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty