Would you like to log in again?
Abdos® Natural Racked Sterile Micro Tips, Graduated
Supplier: Foxx Life Sciences
Abdos®
P10114 P10148 P10147 P10150 P10117
76516-480CS 125.03 USD InStock
76516-480 76516-482 76516-484 76516-486 76516-488
Abdos® Natural Racked Sterile Micro Tips, Graduated
Pipette Tips
Racked Micro Tips are offered in prefilled racks that do not require any manual accumulation in empty boxes. Polypropylene (PP) racks are very robust, sturdy with consistent and thick wall thickness.
  • Racks are stable
  • Can be used for repeat autoclaving
  • Reusable once empty
  • Sterilized by gamma radiation
  • DNase/RNase pyrogen free
Racks are hinged with a transparent lid providing excellent visualization inside the box.
Certifications: ISO Standard: 9001, 13485, 14001, 11137.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty