Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Monoclonal Antibody [clone: M205] (HRP (Horseradish Peroxidase))

Supplier: GenScript Biotech
GenScript®
A01827-200
76198-894EA 120.9 USD InStock
76198-894
Anti-IgG Mouse Monoclonal Antibody [clone: M205] (HRP (Horseradish Peroxidase))
Antibodies
GenScript Anti-Rabbit IgG Antibody (M205) only reacts with native rabbit IgG and does not bind to the denatured or reduced rabbit IgG heavy chain or light chain. The monoclonal antibody has no cross-reactivity with mouse, rat, bovine, goat, or sheep immunoglobulins and limited cross-reactivity with human immunoglobulins.

  • High specificit
  • High affinity
  • High stability

At GenScript, secondary antibodies are purified from the serum of several hosts, including goat, donkey, mouse, rabbit, chicken and sheep. Secondary antibodies are available with specificity for whole Ig molecules or antibody fragments such as the Fc or Fab regions. The conjugated secondary antibodies can be used for various immunology analysis and protein assays.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: M205
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty