Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-V5 Tag Mouse Monoclonal Antibody (iFluor 647) [clone: 4C12E11]

Supplier: Genscript
GenScript®
A01805-100
10498-192EA 364.71 USD InStock
10498-192
Anti-V5 Tag Mouse Monoclonal Antibody (iFluor 647) [clone: 4C12E11]
Antibodies
THE™ V5 tag antibody [iFluor 647], mAb, mouse recognizes N-terminal, internal and C-terminal V5-tagged proteins.

  • Comprehensive pool
  • against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: V5 Tag
Clonality: Monoclonal
Clone: 4C12E11
Conjugation: iFluor 647
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty