Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dibromo-2-nitrobenzene 98%

Supplier: Ambeed
Warning
Synonyms: 2,5-Dibromonitrobenzene

A383588-100G A383588-25G A383588-10G A383588-500G
77251-936EA 5 USD InStock
77251-936 77251-938 76841-114 77251-934
1,4-Dibromo-2-nitrobenzene 98%
1,4-Dibromo-2-nitrobenzene
Caution: for Research Use Only


Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
MDL Number: MFCD00007046
CAS Number: 3460-18-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty