Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iminodiacetic acid 98%

Supplier: Ambeed
Warning
Synonyms: 2,2'-Azanediyldiacetic acid

A617225-100G A617225-500G A617225-1000G A617225-25G
77005-496EA 6.18 USD InStock
77005-496 77005-498 76844-370 77005-494
Iminodiacetic acid 98%
Iminodiacetic acid
Caution: for Research Use Only


Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00004280
CAS Number: 142-73-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty