Would you like to log in again?
Autoclave Bags with Steam Indicator, Self-Seal, Super Duty SMS
Supplier: Keystone Cleanroom Products
BGSMS6SS3636 BGSMS6SS3030 BGSMS6SS09.516 BGSMS6SS1824 BGSMS6SS1520 BGSMS6SS2836 BGSMS6SS1622 BGSMS6SS1217 BGSMS6SS2424 BGSMS6SS2430 BGSMS6SS14.522 BGSMS6SS1318 BGSMS6SS2126 BGSMS6SS2435 BGSMS6SS2028 BGSMS6SS2630 BGSMS6SS1221 BGSMS6SS3648 BGSMS6SS1830 BGSMS6SS4848
76795-438CS 2356.2 USD InStock
76795-438 76795-428 76795-458 76795-436 76795-426 76795-448 76795-456 76795-434 76795-424 76795-446 76795-454 76795-432 76795-444 76795-422 76795-452 76795-430 76795-442 76795-450 76795-440 76795-460
Autoclave Bags with Steam Indicator, Self-Seal, Super Duty SMS
Bags Sterilization Pouches
Self Sealing Super Duty SMS Autoclave Bags from Keystone Cleanroom Products are designed to satisfy the most stringent requirements in the sterilization of critical pharmaceutical equipment and components.
  • Convenient, time saving self-seal closure system
  • Significant improvement versus cutting and wrapping with sheets and tape
  • Superior barrier prevents liquid and microbial penetration
  • Manufactured and packaged in a certified ISO 6 Cleanroom
  • Strong, rip and tear resistant 88 gsm SMS material
  • Sub packed into sealed labeled poly bags with manufacturing lot number and Date of Manufacture for traceability
Designed for higher temperature steam sterilization cycles (135 °C)* and compatible with ethylene oxide and hydrogen peroxide sterilization.
Certifications: Certificates of Conformance sent with every shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty