Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dihydroxybenzoic acid 98%

Supplier: Ambeed
Warning
Synonyms: beta-Resorcylic acid

A105508-100G A105508-10G A105508-25G A105508-500G
77549-008EA 12 USD InStock
77549-008 76869-414 76869-412 77549-010
2,4-Dihydroxybenzoic acid 98%
2,4-Dihydroxybenzoic acid
Caution: for Research Use Only


Formula: C₇H₆O₄
MW: 154.12 g/mol
MDL Number: MFCD00002451
CAS Number: 89-86-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty