Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-1-naphthalenesulfonic acid 98%

Supplier: Ambeed
Warning
Synonyms: 2-Aminonaphthalene-1-sulfonic acid2-Aminonaphthalene-1-sulphonic acid

A149919-25G
76871-580EA 5.15 USD InStock
76871-580
2-Amino-1-naphthalenesulfonic acid 98%
2-Amino-1-naphthalenesulphonic acid
Caution: for Research Use Only


Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00003988
CAS Number: 81-16-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty