Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mannose triflate 95%

Supplier: Ambeed
Warning
Synonyms: TATM(2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose(2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose

A189531-100MG A189531-250MG A189531-5G
76873-012EA 773.53 USD InStock
76873-012 76873-010 76873-008
Mannose triflate 95%
Mannose triflate
Caution: for Research Use Only


Formula: C₁₅H₁₉F₃O₁₂S
MW: 480.36 g/mol
MDL Number: MFCD00012353
CAS Number: 92051-23-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty