Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9,10-Anthraquinone 95%

Supplier: Ambeed
Warning
Synonyms: AnthraquinoneAnthracene-9,10-dione

A259681-100G A259681-500G A259681-1000G
76874-836EA 17.51 USD InStock
76874-836 76874-832 76874-834
9,10-Anthraquinone 95%
9,10-Anthraquinone
Caution: for Research Use Only


Formula: C₁₄H₈O₂
MW: 208.22 g/mol
MDL Number: MFCD00001188
CAS Number: 84-65-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty