Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-4-nitrocinnamate 95%

Supplier: Ambeed
Warning
Synonyms: Ethyl 4-nitrocinnamate

A290818-5G A290818-1G A290818-25G
76875-452EA 93.73 USD InStock
76875-452 76875-450 76875-454
Ethyl-4-nitrocinnamate 95%
Ethyl 4-nitrocinnamate
Caution: for Research Use Only


Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
MDL Number: MFCD00007380
CAS Number: 953-26-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty