Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalacetic acid 97%

Supplier: Ambeed
Danger
Synonyms: Oxalacetic acid2-Succinic acid

A333802-100MG
76876-044EA 5.15 USD InStock
76876-044
Oxalacetic acid 97%
Oxalacetic acid
Caution: for Research Use Only


Formula: C₄H₄O₅
MW: 132.07 g/mol
MDL Number: MFCD00002592
CAS Number: 328-42-7
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty