Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dimethoxynaphthalene 98%

Supplier: Ambeed
Warning

A470491-1G A470491-25G A470491-5G
76877-720EA 76.22 USD InStock
76877-720 76877-718 76877-716
2,3-Dimethoxynaphthalene 98%
2,3-Dimethoxynaphthalene
Caution: for Research Use Only


Formula: C₁₂H₁₂O₂
MDL Number: MFCD05664308
CAS Number: 10103-06-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty