Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-4-chlorophenylacetic acid 98%

Supplier: Ambeed
Danger

A819741-1G
76880-384EA 10.3 USD InStock
76880-384
α-Bromo-4-chlorophenylacetic acid 98%
α-Bromo-4-chlorophenylacetic acid
Caution: for Research Use Only


Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD08276760
CAS Number: 3381-73-5
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty