Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
E.Z.N.A® Total RNA Kit I
Supplier: Omega Bio-Tek
E.Z.N.A®
R6834-03 R6834-03
76539-264EA 1934.13 USD InStock
76539-264 CA76539-264
E.Z.N.A® Total RNA Kit I
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Isolate total RNA from cells or soft tissues using spin column.
  • Rapid – 20 minutes or less
  • Safe – no phenol/chloroform extractions
  • Versatile – spin and vacuum formats available
  • High quality – purified RNA suitable for a variety of downstream applications
The E.Z.N.A.® Total RNA Kit I provides a straightforward and rapid method for the isolation of up to 100µg of total RNA from cultured eukaryotic cells and soft tissues. Multiple samples of up to 1×107 eukaryotic cells or 30mg of tissue can be processed in parallel in fewer than 20 minutes. Purified RNA can be used in many downstream applications such as RT-PCR, Northern blotting, nuclease protection assay, and in vitro translation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty