Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Hyclone™ Ham's F-10 Basal Media, Cell Culture Media, Liquid
Supplier: HyClone products (Cytiva)
HyClone™
SH30025.01 SH30025.01
18777-010EA 36.24 USD InStock
18777-010 CA18777-010
Hyclone™ Ham's F-10 Basal Media, Cell Culture Media, Liquid
Cell Culture Media
Ham's F-10 was designed for use with Chinese Hamster Ovary (CHO) cells, and has been shown to support the growth of human diploid cells, white blood cells, and primary explants from rat, rabbit, or chicken tissues. This classical media can be used for serum-free growth of some cell lines including CHO cells. Serum supplementation may or may not be necessary depending on the type of cell being cultured. Ham's F-10 contains a wide variety of components, including zinc, hypoxanthine, and thymidine and is buffered with sodium bicarbonate.
  • Contains 6,1 mM glucose
  • All liquid media products are hydrated using WFI quality water
  • 0,1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes
A Certificate of Analysis is included in each package with preparation instructions, intended use, and handling and performance criteria.
Ordering information: Liquid media are supplied in unbreakable, square PETG bottles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty