Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffered oxide etch 5:1 modified, Electronic Grade, Puritan Products™

Supplier: Puritan Products, Inc.
Danger

330575
76551-218CS 385 USD InStock
76551-218
Buffered oxide etch 5:1 modified, Electronic Grade, Puritan Products™
Buffered oxide etch


UN: 2922
ADR: 8, 6.1,II
Order Now
Specification Test Results
Ammonium Fluoride (wt%) 29.82-30.82%
Assay (w/w) 7.62-7.82%
Aluminum 0.2 ppm maximum
Arsenic and Antimony 0.03 ppm maximum
Boron 0.2 ppm maximum
Calcium 0.2 ppm maximum
Chromium 0.1 ppm maximum
Copper 0.1 ppm maximum
Iron 0.2 ppm maximum
Lead 0.3 ppm maximum
Magnesium 0.2 ppm maximum
Manganese 0.2 ppm maximum
Nickel 0.2 ppm maximum
Potassium 0.3 ppm maximum
Sodium 0.3 ppm maximum
Tin 0.3 ppm maximum
Titanium 0.3 ppm maximum
Zinc 0.3 ppm maximum
Tested to the current release of Semi Book of Standards Process Chemicals

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty