Would you like to log in again?
AccuForge® Double Edge Blades, Unwrapped
Supplier: AccuTec Blades
AccuForge®
AGBL-6001-0000
76717-292CS 1029.8 USD InStock
76717-292
AccuForge® Double Edge Blades, Unwrapped
Razors and Blades
AccuForge double edge blades feature a 3-Facet edge for exacting, clean cuts. The MicroCoat® Enrichment technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. No additional oil applied to blades.
  • Blade edge with MicroCoat® technology enhances durability and sharpness
  • 3-facet edge for increased sharpness
  • Blades are supplied on a bayonet
  • Bulk unwrapped blades are ideal for high volume uses
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty