Would you like to log in again?
miniPCR® Sickle Cell Genetics Lab: Diagnosing Baby Marie™
Supplier: AMPLYUS LLC
miniPCR®
KT-1502-01
470352-006EA 75.19 USD InStock
470352-006
miniPCR® Sickle Cell Genetics Lab: Diagnosing Baby Marie™
Educational Classroom Kits and Activities
Baby Marie Robinson needs to be tested for sickle cell disease.
  • Techniques: Micropipetting, gel electrophoresis
  • Topics: Genetic inheritance, genes and disease, genotype to phenotype, protein structure and function, natural selection, molecular diagnostics, biotechnology
  • Time required: Can be completed in one 45-minute class period
  • Level: General high school through college
In this engaging case study, students test a fictional family for sickle cell disease. The Robinson family has two children, one of whom has had an initial test result indicating possible sickle cell disease. Students will use gel electrophoresis to test the Robinson family for the sickle cell allele and give baby Marie Robinson a definitive diagnosis.
Reagents and consumables supplied by user:
- Agarose (electrophoresis grade)
- DNA stain (e.g., GelGreen™)
- Gel electrophoresis buffer (e.g., 1X TBE)
- Distilled or deionized H₂O (to dilute buffer concentrate – not included in Lab Companion Kit)
- Microtubes
Required equipment:
- Micropipettes and tips
- Gel electrophoresis and fluorescence visualization system
Storage notes:
- Reagents require freezer storage
- Reagents are stable for 12 months when stored in the freezer
Accessories information: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery information: The Sickle Cell Genetics Lab kit contains reagents for 8 lab groups: 4 DNA samples; 1 tube of DNA molecular weight marker (DNA Ladder). Note: Reagents are compatible with blueGel™ or other standard horizontal electrophoresis systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty