Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyperSep™ Retain AX SPE Plates

Supplier: Thermo Scientific
HyperSep™
60303-408
10048-500EA 606.58 USD InStock
10048-500
HyperSep™ Retain AX SPE Plates
Microplates Solid Phase Extraction Plates (SPE)
Versatile polymeric material for retention of acidic compounds.

  • Particle size: 40 to 60 µm
  • Pore size: 55 to 90 Å
  • Surface area: 550 to 750 m²/g

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty