Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 5% USP (Hypertonic)

Supplier: B BRAUN MEDICAL
Synonyms: HaliteSea saltSaline

L8061
76313-498CS 455.08 USD InStock
76313-498
Sodium chloride 5% USP (Hypertonic)
Sodium chloride
5% Sodium Chloride Injection USP are sterile, nonpyrogenic and contain no bacteriostatic or antimicrobial agents. This intravenous solution is indicated for use in adults and pediatric patients as source of electrolytes and water for hydration.

  • NDC number: 00264-7806-10

Not manufactured with latex, PVC or DEHP

Specific Gravity: 1.021


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty