Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% USP for injection

Supplier: B BRAUN MEDICAL
Synonyms: HaliteSea saltSaline

L8002
95037-774CS 277.12 USD InStock
95037-774
Sodium chloride 0.9% USP for injection
Sodium chloride
0.9% Sodium Chloride Injection USP is indicated for extracellular fluid replacement, treatment of metabolic alkalosis in the presence of fluid loss and mild sodium depletion.

Not manufactured with latex, PVC or DEHP.

Specific Gravity: 1.006

NDC number: 0264-7800-20


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty