Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% USP for injection

Supplier: B BRAUN MEDICAL
Synonyms: HaliteSea saltSaline

S8004-5264
76363-600CS 424.21 USD InStock
76363-600
Sodium chloride 0.9% USP for injection
Sodium chloride
Sodium Chloride Injections USP are sterile, nonpyrogenic, isotonic and contain no bacteriostatic or antimicrobial agents. These intravenous solutions are indicated for use in adults and pediatric patients as sources of electrolytes and water for hydration.

Not manufactured with latex, PVC or DEHP.

NDC number: 00264-1800-32

Shelf life: 15 months (total)


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty