Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% USP

Supplier: Fresenius Kabi
Synonyms: HaliteSea saltSaline

924830
68099-166PK 111.06 USD InStock
68099-166
Sodium chloride 0.9% USP
Sodium chloride
Sterile injections comply with USP standards and are bar coded for maximum traceability.

  • Preservative-free
  • NDC number: 924-30


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty