Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% USP for irrigation

Supplier: BAXTER IV SOLUTIONS
Synonyms: HaliteSea saltSaline

2B7479
68000-520CS 253.14 USD InStock
68000-520
Sodium chloride 0.9% USP for irrigation
Sodium chloride
Solutions for irrigation (for Preparation of Slushed Solution)

  • mEq/L: Na 154, Cl 154
  • mOsmol/L (calc): 308
  • NDC Number: 0338-0047-29
  • pH: 5.5 (4.5–7.0)


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty