Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dichloro-6-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

17317-25G 17317-5G
76883-692EA 301.79 USD InStock
76883-692 76883-694
2,3-Dichloro-6-(trifluoromethyl)benzoic acid ≥97% research grade
2,3-Dichloro-6-(trifluoromethyl)benzoic acid
Caution: for Research Use Only


Formula: C₈H₃Cl₂F₃O₂
MW: 259.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660270
CAS Number: 25922-43-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty