Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethoxy-2,6-difluorophenol ≥95%

Supplier: AOBChem USA

Warning

22758-1G 22758-25G 22758-5G
76910-954EA 645.81 USD InStock
76910-954 76910-956 76910-952
3-Ethoxy-2,6-difluorophenol ≥95%
3-Ethoxy-2,6-difluorophenol
Caution: for Research Use Only


Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11519373
CAS Number: 1092461-31-8
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty