Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxy-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

26430-1G 26430-25G 26430-5G
76912-380EA 379.04 USD InStock
76912-380 76912-376 76912-378
3-Methoxy-4-methylphenol ≥97%
3-Methoxy-4-methylphenol
Caution: for Research Use Only


Formula: C₈H₁₀O₂
MW: 138.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16998731
CAS Number: 19217-50-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty