Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyloxy-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning
Synonyms: 3-Methoxy-4-benzyloxybenzyl alcohol

29192-5G 29192-25G
76913-194EA 60.77 USD InStock
76913-194 76913-192
4-Benzyloxy-3-methoxybenzyl alcohol ≥97%
4-Benzyloxy-3-methoxybenzyl alcohol
Caution: for Research Use Only


Formula: C₁₅H₁₆O₃
MW: 244.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004640
CAS Number: 33693-48-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty