Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

10200-25G
76894-294EA 109.18 USD InStock
76894-294
2-Chloro-4-(trifluoromethyl)phenylboronic acid ≥97% research grade
2-Chloro-4-(trifluoromethyl)phenylboronic acid
Caution: for Research Use Only


Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684337
CAS Number: 254993-59-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty