Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-1-chloro-3,5-difluorobenzene ≥95% research grade

Supplier: AOBChem USA

Warning

47700-5G 47700-25G 47700-1G
76904-186EA 85.49 USD InStock
76904-186 76904-188 76904-190
2-Bromo-1-chloro-3,5-difluorobenzene ≥95% research grade
2-Bromo-1-chloro-3,5-difluorobenzene


Formula: C₆H₂BrClF₂
MW: 227.434 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11849939
CAS Number: 1020198-58-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty