Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-bromo-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54995-25G
76905-112EA 144.2 USD InStock
76905-112
2-Amino-3-bromo-5-methylbenzoic acid ≥97%
2-Amino-3-bromo-5-methylbenzoic acid
Caution: for Research Use Only


Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00051705
CAS Number: 13091-43-5
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty