Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-(trifluoromethyl)anisole ≥97%

Supplier: AOBChem USA

Warning

27004-5G 27004-25G
76929-372EA 216.3 USD InStock
76929-372 76929-374
4-Chloro-3-(trifluoromethyl)anisole ≥97%
4-Chloro-3-(trifluoromethyl)anisole
Caution: for Research Use Only


Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631404
CAS Number: 400-73-7
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty