Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-3-iodophenol ≥95% research grade

Supplier: AOBChem USA

Warning

27117-1G 27117-25G 27117-500MG 27117-5G
76929-422EA 140.08 USD InStock
76929-422 76929-424 76929-426 76929-428
4-Bromo-3-iodophenol ≥95% research grade
4-Bromo-3-iodophenol
Caution: for Research Use Only


Formula: C₆H₄BrIO
MW: 298.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070745
CAS Number: 202865-84-7
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty