Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoroacetyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning
Synonyms: 4-(2,2,2-trifluoroacetyl)benzoic acid

107413-5G 107413-25G
76906-474EA 96.82 USD InStock
76906-474 76906-472
4-(Trifluoroacetyl)benzoic acid ≥95%
4-(Trifluoroacetyl)benzoic acid
Caution: for Research Use Only


Formula: C₉H₅F₃O₃
MW: 218.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052340
CAS Number: 58808-59-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty