Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-4-(methoxymethoxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14002-1G 14002-5G 14002-25G
76907-308EA 989.83 USD InStock
76907-308 76907-306 76907-310
3-Fluoro-4-(methoxymethoxy)phenylboronic acid ≥97% research grade
3-Fluoro-4-(methoxymethoxy)phenylboronic acid


Formula: C₈H₁₀BFO₄
MW: 199,972 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617248
CAS Number: 1315341-79-7
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty