Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning
Synonyms: 3-Iodoindazole

25779-25G
76912-052EA 75.19 USD InStock
76912-052
3-Iodo-1H-indazole ≥97%
3-Iodo-1H-indazole
Caution: for Research Use Only


Formula: C₇H₅IN₂
MW: 244.0325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972469
CAS Number: 66607-27-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty