Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethoxy)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

52976-25G
76916-926EA 136.99 USD InStock
76916-926
3-(Difluoromethoxy)benzaldehyde ≥97% research grade
3-(Difluoromethoxy)benzaldehyde
Caution: for Research Use Only


Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236222
CAS Number: 85684-61-3
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty