Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning
Synonyms: 6-Bromopyridine-3-boronic acid6-Bromo-3-pyridinylboronic acid2-Bromo-5-pyridinylboronic acid2-Bromopyridine-5-boronic acid(6-Bromopyridin-3-yl)boronic acid

10158-25G
76894-270EA 60.77 USD InStock
76894-270
2-Bromo-5-pyridineboronic acid ≥97%
2-Bromo-5-pyridineboronic acid
Caution: for Research Use Only


Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411558
CAS Number: 223463-14-7
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty